Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC19
Entrez Gene ID
100140471
Gene Name
leucine rich repeat containing 19
Gene Aliases
-
Chromosome Location
Chr.8: 17242194 - 17257138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191267.1 NP_001178196.1 1 - 2 86 100

View Full Product Details