Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAX3
Entrez Gene ID
540951
Gene Name
paired box 3
Gene Aliases
-
Chromosome Location
Chr.2: 110369553 - 110471285 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206818.2 NP_001193747.1 6 - 7 1238 87

View Full Product Details