Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K9
Entrez Gene ID
538340
Gene Name
mitogen-activated protein kinase kinase kinase 9
Gene Aliases
-
Chromosome Location
Chr.10: 82368180 - 82454886 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206852.1 NP_001193781.1 10 - 11 2019 113

View Full Product Details