Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDH12
Entrez Gene ID
540672
Gene Name
cadherin 12
Gene Aliases
-
Chromosome Location
Chr.20: 51192127 - 51567420 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206958.1 NP_001193887.1 8 - 9 1516 59

View Full Product Details