Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC20B
Entrez Gene ID
522049
Gene Name
cell division cycle 20B
Gene Aliases
-
Chromosome Location
Chr.20: 23940884 - 24005261 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206982.1 NP_001193911.1 4 - 5 641 69

View Full Product Details