Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPLX4
Entrez Gene ID
616684
Gene Name
complexin 4
Gene Aliases
-
Chromosome Location
Chr.24: 58263769 - 58286282 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191350.1 NP_001178279.1 2 - 3 257 90

View Full Product Details