Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAP30L
Entrez Gene ID
533462
Gene Name
SAP30 like
Gene Aliases
-
Chromosome Location
Chr.7: 65722279 - 65735762 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191372.3 NP_001178301.1 2 - 3 642 75

View Full Product Details