Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PABPC1
Entrez Gene ID
282296
Gene Name
poly(A) binding protein cytoplasmic 1
Gene Aliases
PABP-1
Chromosome Location
Chr.14: 63625778 - 63643551 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174568.3 NP_776993.1 9 - 10 1847 99
GenBank mRNA BC102365.1 - 9 - 10 1837 99
EE252871.1 - 2 - 3 332 99

View Full Product Details