Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALR1
Entrez Gene ID
539429
Gene Name
galanin receptor 1
Gene Aliases
-
Chromosome Location
Chr.24: 2145784 - 2156850 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192241.1 NP_001179170.1 1 - 2 663 82

View Full Product Details