Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC36
Entrez Gene ID
512403
Gene Name
leucine rich repeat containing 36
Gene Aliases
-
Chromosome Location
Chr.18: 34936744 - 35001719 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192321.1 NP_001179250.1 1 - 2 68 87

View Full Product Details