Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDH8
Entrez Gene ID
513346
Gene Name
protocadherin 8
Gene Aliases
-
Chromosome Location
Chr.12: 10909007 - 10914329 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192348.3 NP_001179277.2 2 - 3 2834 88

View Full Product Details