Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTDSP1
Entrez Gene ID
516199
Gene Name
CTD small phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.2: 106408226 - 106417344 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192440.1 NP_001179369.1 4 - 5 483 88

View Full Product Details