Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHYKPL
Entrez Gene ID
537241
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
Gene Aliases
AGXT2L2
Chromosome Location
Chr.7: 39286822 - 39312379 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192899.2 NP_001179828.1 3 - 4 338 73

View Full Product Details