Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALHM3
Entrez Gene ID
613539
Gene Name
calcium homeostasis modulator 3
Gene Aliases
-
Chromosome Location
Chr.26: 24059298 - 24065042 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192960.2 NP_001179889.2 2 - 3 548 63

View Full Product Details