Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB1
Entrez Gene ID
526903
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 76649127 - 76679517 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193001.1 NP_001179930.1 1 - 2 215 58
GenBank mRNA BC151724.1 - 1 - 2 213 58

View Full Product Details