Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC32
Entrez Gene ID
615467
Gene Name
leucine rich repeat containing 32
Gene Aliases
-
Chromosome Location
Chr.15: 56043489 - 56057577 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193075.2 NP_001180004.1 1 - 2 105 109

View Full Product Details