Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRG1
Entrez Gene ID
615508
Gene Name
tumor protein p63 regulated 1
Gene Aliases
-
Chromosome Location
Chr.1: 78014339 - 78169293 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193079.1 NP_001180008.1 3 - 4 544 81

View Full Product Details