Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R3F
Entrez Gene ID
513859
Gene Name
protein phosphatase 1 regulatory subunit 3F
Gene Aliases
-
Chromosome Location
Chr.X: 87307832 - 87324338 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193144.1 NP_001180073.1 1 - 2 986 87

View Full Product Details