Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNC93B1
Entrez Gene ID
508878
Gene Name
unc-93 homolog B1, TLR signaling regulator
Gene Aliases
-
Chromosome Location
Chr.29: 45516985 - 45527085 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193147.1 NP_001180076.1 1 - 2 122 100

View Full Product Details