Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIS3
Entrez Gene ID
617311
Gene Name
Meis homeobox 3
Gene Aliases
MEIS3P1
Chromosome Location
Chr.18: 54386162 - 54397195 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193152.2 NP_001180081.1 7 - 8 816 65

View Full Product Details