Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAC8L1
Entrez Gene ID
618331
Gene Name
PLAC8 like 1
Gene Aliases
-
Chromosome Location
Chr.7: 57296638 - 57317430 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193185.1 NP_001180114.1 1 - 2 123 99

View Full Product Details