Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK8
Entrez Gene ID
618438
Gene Name
kallikrein related peptidase 8
Gene Aliases
KLNH
Chromosome Location
Chr.18: 56972257 - 56978001 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193189.1 NP_001180118.1 2 - 3 237 95

View Full Product Details