Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBLN1
Entrez Gene ID
618604
Gene Name
cerebellin 1 precursor
Gene Aliases
-
Chromosome Location
Chr.18: 17738055 - 17740658 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195052.1 NP_001181981.1 1 - 2 266 82

View Full Product Details