Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM258
Entrez Gene ID
767958
Gene Name
transmembrane protein 258
Gene Aliases
C11orf10, C29H11orf10
Chromosome Location
Chr.29: 40232832 - 40236557 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195063.1 NP_001181992.1 2 - 3 200 79
GenBank mRNA BC108218.1 - 2 - 3 173 79

View Full Product Details