Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
RAB1B
Entrez Gene ID
540685
Gene Name
RAB1B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.29: 44324272 - 44331570 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076233.1 NP_001069701.1 3 - 4 237 83
GenBank mRNA BC105393.1 - 3 - 4 237 83

View Full Product Details