Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GID4
Entrez Gene ID
509503
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C19H17orf39
Chromosome Location
Chr.19: 34509801 - 34524072 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199762.1 NP_001186691.1 3 - 4 564 97
GenBank mRNA BC102715.1 - 3 - 4 428 97
DT807444.1 - 3 - 4 347 97
DT829639.1 - 3 - 4 428 97
EE341469.1 - 1 - 2 40 97

View Full Product Details