Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM71E1
Entrez Gene ID
617892
Gene Name
family with sequence similarity 71 member E1
Gene Aliases
-
Chromosome Location
Chr.18: 56607980 - 56613553 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205375.2 NP_001192304.1 2 - 3 209 76
GenBank mRNA BC109767.1 - 2 - 3 210 76

View Full Product Details