Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PPARG-TSEN2
- Entrez Gene ID
-
724061
- Gene Name
- PPARG-TSEN2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 56618560 - 56832975 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003097.1 |
|
8 - 9 |
1435 |
96 |
GenBank mRNA |
DQ256760.1 |
- |
8 - 9 |
1435 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
peroxisome proliferator activated receptor gamma |
PPARG |
|
|
Sense |
Chr. - 56710574 - 56775806 on Build ARS-UCD1.2 |
Downstream |
microRNA mir-2373 |
MIR2373 |
bta-mir-2373 |
104.2kb
|
Anti-sense |
Chr. - 56833146 - 56833223 on Build ARS-UCD1.2 |
View Full Product Details