Product Details

SNP ID
rs77850066
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:116711651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCATTAAAGATAGCTGCCATT[A/G]CTAATGATCAAGCACAGTTCAATGA
Phenotype
MIM: 616057
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TUSC7 PubMed Links

Gene Details

Gene
TUSC7
Gene Name
tumor suppressor candidate 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details