Product Details

SNP ID
rs75472800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129004718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCCTTGAGCTAAGTACTTTA[C/T]GTGGCTTAACTCATTTAAGCCTCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFCC1 PubMed Links

Gene Details

Gene
EFCC1
Gene Name
EF-hand and coiled-coil domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024768.2 Intron NP_079044.2
XM_005247781.3 Intron XP_005247838.1
XM_011513161.2 Intron XP_011511463.1
XM_011513163.2 Intron XP_011511465.1
XM_011513164.2 Intron XP_011511466.1
Gene
KIAA1257
Gene Name
KIAA1257
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020741.2 Intron NP_065792.1
XM_011513050.2 Intron XP_011511352.2
XM_017006939.1 Intron XP_016862428.1
XM_017006940.1 Intron XP_016862429.1
XM_017006941.1 Intron XP_016862430.1
XM_017006942.1 Intron XP_016862431.1
XM_017006943.1 Intron XP_016862432.1
XM_017006944.1 Intron XP_016862433.1
XM_017006945.1 Intron XP_016862434.1
XM_017006946.1 Intron XP_016862435.1
XM_017006947.1 Intron XP_016862436.1
XM_017006948.1 Intron XP_016862437.1
XM_017006949.1 Intron XP_016862438.1
XM_017006950.1 Intron XP_016862439.1
XM_017006951.1 Intron XP_016862440.1
XM_017006952.1 Intron XP_016862441.1
XM_017006953.1 Intron XP_016862442.1
XM_017006954.1 Intron XP_016862443.1

View Full Product Details