Product Details

SNP ID
rs79839148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138951595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCAAGTGAGGCCCTCTTCCCT[A/C]CAAAGACCCGTGGGATGCTCTCAGA
Phenotype
MIM: 605597
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOXL2 PubMed Links

Gene Details

Gene
FOXL2
Gene Name
forkhead box L2
There are no transcripts associated with this gene.

Gene
FOXL2NB
Gene Name
FOXL2 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040061.2 1682 UTR 3 NP_001035150.1
XM_005247443.3 1682 UTR 3 XP_005247500.1
Gene
LINC01391
Gene Name
long intergenic non-protein coding RNA 1391
There are no transcripts associated with this gene.

View Full Product Details