Product Details

SNP ID
rs74346735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98734496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCTGTTCTTGTGGTCATGTAGA[C/T]TGGGAGGGGAGATCTTCATTCCTTG
Phenotype
MIM: 607156
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST3GAL6 PubMed Links

Gene Details

Gene
ST3GAL6
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271142.1 Intron NP_001258071.1
NM_001271145.1 Intron NP_001258074.1
NM_001271146.1 Intron NP_001258075.1
NM_001271147.1 Intron NP_001258076.1
NM_001271148.1 Intron NP_001258077.1
NM_001323352.1 Intron NP_001310281.1
NM_001323353.1 Intron NP_001310282.1
NM_001323358.1 Intron NP_001310287.1
NM_001323359.1 Intron NP_001310288.1
NM_001323360.1 Intron NP_001310289.1
NM_001323362.1 Intron NP_001310291.1
NM_001323363.1 Intron NP_001310292.1
NM_001323364.1 Intron NP_001310293.1
NM_001323365.1 Intron NP_001310294.1
NM_001323366.1 Intron NP_001310295.1
NM_001323367.1 Intron NP_001310296.1
NM_001323368.1 Intron NP_001310297.1
NM_006100.3 Intron NP_006091.1
Gene
ST3GAL6-AS1
Gene Name
ST3GAL6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details