Product Details

SNP ID
rs79276520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151327133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGATGCAGTCAAGAATATAGCTA[A/T]CTAGAAAAGGGAGATCTGCATTGAG
Phenotype
MIM: 611318 MIM: 600515 MIM: 606380
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MED12L PubMed Links

Gene Details

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 1944 Intron NP_443728.3
XM_006713487.3 1944 Intron XP_006713550.1
XM_011512390.2 1944 Intron XP_011510692.1
XM_011512394.2 1944 Intron XP_011510696.1
XM_011512399.2 1944 Intron XP_011510701.1
XM_017005676.1 1944 Intron XP_016861165.1
XM_017005677.1 1944 Intron XP_016861166.1
XM_017005678.1 1944 Intron XP_016861167.1
XM_017005679.1 1944 Intron XP_016861168.1
XM_017005680.1 1944 Intron XP_016861169.1
XM_017005681.1 1944 Intron XP_016861170.1
Gene
P2RY12
Gene Name
purinergic receptor P2Y12
There are no transcripts associated with this gene.

Gene
P2RY13
Gene Name
purinergic receptor P2Y13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176894.2 1944 UTR 3 NP_795713.2
XM_006713664.1 1944 UTR 3 XP_006713727.1

View Full Product Details