Product Details

SNP ID
rs78466645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160262778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCTTGTCTTTAGCTCAAAAAT[A/G]TAAGTGTCAAAGAGACATATTTTGG
Phenotype
MIM: 611177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT80 PubMed Links

Gene Details

Gene
IFT80
Gene Name
intraflagellar transport 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190241.1 Intron NP_001177170.1
NM_001190242.1 Intron NP_001177171.1
NM_020800.2 Intron NP_065851.1

View Full Product Details