Product Details

SNP ID
rs79136918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179151652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAAAGCCCTGGCTGTCAAGGAC[C/T]TGTATATTTGGTCCTTGCTTCTTTC
Phenotype
MIM: 171834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928739 PubMed Links

Gene Details

Gene
LOC101928739
Gene Name
uncharacterized LOC101928739
There are no transcripts associated with this gene.

Gene
PIK3CA
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006218.3 Intron NP_006209.2
XM_006713658.3 Intron XP_006713721.1
XM_011512894.2 Intron XP_011511196.1

View Full Product Details