Product Details

SNP ID
rs79858408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48491220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGAGAGGGGCCGTCGCCACGGA[A/G]CCCGGAGCTGGGCAAGGAGCGGCTG
Phenotype
MIM: 607520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRYL PubMed Links

Gene Details

Gene
FRYL
Gene Name
FRY like transcription coactivator
There are no transcripts associated with this gene.

Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
There are no transcripts associated with this gene.

Gene
ZAR1
Gene Name
zygote arrest 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175619.2 969 Missense Mutation AAC,AGC N310S NP_783318.1

View Full Product Details