Product Details
- SNP ID
-
rs75488359
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:2713606 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTTTTCCCCTTAGGTTTTCCATC[A/G]TTTATCAAGATCATTCCTAAGAACT
- Phenotype
-
MIM: 606198
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IRX2
PubMed Links
Gene Details
- Gene
- IRX2
- Gene Name
- iroquois homeobox 2
View Full Product Details