Product Details
- SNP ID
-
rs75246236
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:34845500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATACCTACAGGTCACAGCAAACC[A/G]AGATCCCAGCTCACCAGCAAACAGA
- Phenotype
-
MIM: 616344
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTC23L
PubMed Links
Gene Details
- Gene
- TTC23L
- Gene Name
- tetratricopeptide repeat domain 23 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317949.1 |
366 |
Missense Mutation |
AAG,GAG |
K37E |
NP_001304878.1 |
NM_144725.3 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
NP_653326.3 |
XM_011513984.2 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_011512286.1 |
XM_011513985.2 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_011512287.1 |
XM_011513987.2 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_011512289.1 |
XM_017009119.1 |
366 |
Missense Mutation |
AAG,GAG |
K37E |
XP_016864608.1 |
XM_017009120.1 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_016864609.1 |
XM_017009121.1 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_016864610.1 |
XM_017009122.1 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_016864611.1 |
XM_017009123.1 |
366 |
Missense Mutation |
AAG,GAG |
K28E |
XP_016864612.1 |
XM_017009124.1 |
366 |
Intron |
|
|
XP_016864613.1 |
XM_017009125.1 |
366 |
Intron |
|
|
XP_016864614.1 |
XM_017009126.1 |
366 |
Intron |
|
|
XP_016864615.1 |
View Full Product Details