Product Details

SNP ID
rs75808692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:445247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCTGAGCACCTGTGGTCTGCA[C/T]GGGGCCTGCATAAAGGAGGGAAGCA
Phenotype
MIM: 606517 MIM: 608186
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHRR PubMed Links

Gene Details

Gene
AHRR
Gene Name
aryl-hydrocarbon receptor repressor
There are no transcripts associated with this gene.

Gene
EXOC3
Gene Name
exocyst complex component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007277.4 Intron NP_009208.2
XM_011513948.2 Intron XP_011512250.1
XM_011513949.2 Intron XP_011512251.1
XM_011513950.2 Intron XP_011512252.1
XM_011513951.2 Intron XP_011512253.1
XM_017009000.1 Intron XP_016864489.1
XM_017009001.1 Intron XP_016864490.1
XM_017009002.1 Intron XP_016864491.1
XM_017009003.1 Intron XP_016864492.1
XM_017009004.1 Intron XP_016864493.1
XM_017009005.1 Intron XP_016864494.1
XM_017009006.1 Intron XP_016864495.1
Gene
EXOC3-AS1
Gene Name
EXOC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details