Product Details
- SNP ID
-
rs74618835
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:79071359 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTTCTGATCACCTCCTACTACA[C/T]AGTGAAGTTGATCCCCACTGAGTAT
- Phenotype
-
MIM: 605932
MIM: 605849
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BHMT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60966081] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BHMT2
- Gene Name
- betaine--homocysteine S-methyltransferase 2
- Gene
- DMGDH
- Gene Name
- dimethylglycine dehydrogenase
There are no transcripts associated with this gene.
View Full Product Details