Product Details

SNP ID
rs77855786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80557735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATATGGAGAAAGAATAATTCCGT[A/G]AGATAACTTTACAGGTAAGTCTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD34B PubMed Links

Gene Details

Gene
ANKRD34B
Gene Name
ankyrin repeat domain 34B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004441.2 2715 UTR 3 NP_001004441.2
XM_006714599.3 2715 UTR 3 XP_006714662.1
XM_011543366.2 2715 UTR 3 XP_011541668.1
XM_011543369.2 2715 UTR 3 XP_011541671.1

View Full Product Details