Product Details

SNP ID
rs77994016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:84038049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGTGCTTCTTGCATTTGGGGCT[C/T]ACACAGGGGCCTAGGTGGCTGGATC
Phenotype
MIM: 615410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRAP2 PubMed Links

Gene Details

Gene
MRAP2
Gene Name
melanocortin 2 receptor accessory protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138409.2 Intron NP_612418.2
XM_011535400.2 Intron XP_011533702.1
XM_017010219.1 Intron XP_016865708.1
XM_017010220.1 Intron XP_016865709.1
XM_017010221.1 Intron XP_016865710.1

View Full Product Details