Product Details

SNP ID
rs77665915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10432963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTCAGCAGCAAATTTACAATCA[A/C]TCTTATTTTCCAGTGTCTCCAGAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00518 PubMed Links

Gene Details

Gene
LINC00518
Gene Name
long intergenic non-protein coding RNA 518
There are no transcripts associated with this gene.

Gene
MIR5689
Gene Name
microRNA 5689
There are no transcripts associated with this gene.

Gene
MIR5689HG
Gene Name
MIR5689 host gene
There are no transcripts associated with this gene.

View Full Product Details