Product Details

SNP ID
rs77288568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10697063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTACTGTGTTCTAGAGTGATGA[C/T]ACCCAGTGTTGCCTGCCCACCTCCT
Phenotype
MIM: 607811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C6orf52 PubMed Links

Gene Details

Gene
C6orf52
Gene Name
chromosome 6 open reading frame 52
There are no transcripts associated with this gene.

Gene
PAK1IP1
Gene Name
PAK1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017906.2 Intron NP_060376.2
XM_005249204.2 Intron XP_005249261.1
XM_011514720.1 Intron XP_011513022.1
XM_011514721.1 Intron XP_011513023.1

View Full Product Details