Product Details

SNP ID
rs79949269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28243634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATCAGATACCTAGGATTCCCA[A/G]TGTGCTTCTGAACAGACACATTTTC
Phenotype
MIM: 611643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN4 PubMed Links

Gene Details

Gene
ZKSCAN4
Gene Name
zinc finger with KRAB and SCAN domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304506.1 4131 UTR 3 NP_001291435.1
NM_019110.4 4131 UTR 3 NP_061983.2
XM_005249095.3 4131 UTR 3 XP_005249152.1
XM_005249097.3 4131 UTR 3 XP_005249154.1
XM_005249098.3 4131 UTR 3 XP_005249155.1
XM_005249100.4 4131 UTR 3 XP_005249157.2
XM_006715095.3 4131 UTR 3 XP_006715158.1
XM_011514588.2 4131 UTR 3 XP_011512890.1
XM_011514589.2 4131 UTR 3 XP_011512891.1
XM_017010844.1 4131 UTR 3 XP_016866333.1

View Full Product Details