Product Details

SNP ID
rs77374931
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4713486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGAGGTGACTAAAGACAACTC[A/G]ACTCTATCTGTATTATTTGAATTTC
Phenotype
MIM: 603778
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CDYL PubMed Links

Gene Details

Gene
CDYL
Gene Name
chromodomain protein, Y-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143970.1 Intron NP_001137442.1
NM_001143971.1 Intron NP_001137443.1
NM_004824.3 Intron NP_004815.3
XM_011514997.2 Intron XP_011513299.1

View Full Product Details