Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039459.2 | 2355 | UTR 3 | NP_001034548.1 | ||
NM_001206480.2 | 2355 | UTR 3 | NP_001193409.1 | ||
NM_001206482.1 | 2355 | UTR 3 | NP_001193411.1 | ||
NM_014800.10 | 2355 | UTR 3 | NP_055615.8 | ||
NM_130442.3 | 2355 | UTR 3 | NP_569709.1 | ||
XM_005249919.2 | 2355 | Intron | XP_005249976.1 | ||
XM_006715805.1 | 2355 | Intron | XP_006715868.1 | ||
XM_011515654.1 | 2355 | Intron | XP_011513956.1 | ||
XM_017012838.1 | 2355 | Intron | XP_016868327.1 | ||
XM_017012839.1 | 2355 | Intron | XP_016868328.1 |