Product Details

SNP ID
rs79101283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6497110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTACCTTGACCACACCTATT[C/T]CACTCCCAGGCTCCCCACTTCCAAT
Phenotype
MIM: 610639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRID2IP PubMed Links

Gene Details

Gene
GRID2IP
Gene Name
Grid2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145118.1 4300 UTR 3 NP_001138590.1
XM_011515404.2 4300 UTR 3 XP_011513706.1

View Full Product Details