Product Details

SNP ID
rs74779554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141919511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTTCTCTTTGGTTCTTTGAT[C/T]CCTACAATTGTCTCCAACGCCTACA
Phenotype
MIM: 604987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLEC5A PubMed Links

Gene Details

Gene
CLEC5A
Gene Name
C-type lectin domain family 5 member A
There are no transcripts associated with this gene.

Gene
OR9A4
Gene Name
olfactory receptor family 9 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001656.1 636 Silent Mutation ATC,ATT I212I NP_001001656.1

View Full Product Details