Product Details

SNP ID
rs75285465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76328065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTGTGGTGTCATTACACGTCTC[A/G]GGAGTTCCTCGTCACTGACTTTATA
Phenotype
MIM: 605356
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YWHAG PubMed Links

Gene Details

Gene
YWHAG
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012479.3 2473 UTR 3 NP_036611.2

View Full Product Details